Assay Details
Gene Symbol: Cct6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033684.1 | NP_001028856.1 | 2 - 3 | 221 | 103 | |
GenBank mRNA | BC104703.1 | - | 2 - 3 | 221 | 103 | |
FQ216833.1 | - | 2 - 3 | 239 | 103 | ||
FQ218432.1 | - | 2 - 3 | 268 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
