Assay Details
Gene Symbol: Cct6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033684.1 | NP_001028856.1 | 7 - 8 | 893 | 91 | |
GenBank mRNA | BC104703.1 | - | 7 - 8 | 893 | 91 | |
FQ216833.1 | - | 7 - 8 | 911 | 91 | ||
FQ218432.1 | - | 7 - 8 | 940 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
