Assay Details
Gene Symbol: Rsrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014128.1 | NP_001014150.1 | 3 - 4 | 353 | 65 | |
XM_006249338.2 | XP_006249400.1 | 2 - 3 | 319 | 65 | ||
XM_006249339.2 | XP_006249401.1 | 2 - 3 | 319 | 65 | ||
XM_006249340.2 | XP_006249402.1 | 2 - 3 | 311 | 65 | ||
XM_006249341.2 | XP_006249403.1 | 2 - 3 | 311 | 65 | ||
GenBank mRNA | BC087067.1 | - | 3 - 4 | 353 | 65 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC087067.1, NM_001014128.1, XM_006249340.2, XM_006249341.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
