Assay Details
Gene Symbol: Rsrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014128.1 | NP_001014150.1 | 6 - 7 | 107 | ||
XM_006249338.2 | XP_006249400.1 | 4 - 5 | 107 | |||
XM_006249339.2 | XP_006249401.1 | 4 - 5 | 107 | |||
XM_006249340.2 | XP_006249402.1 | 5 - 6 | 107 | |||
XM_006249341.2 | XP_006249403.1 | 5 - 6 | 107 | |||
GenBank mRNA | BC087067.1 | - | 6 - 7 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
