Assay Details
Gene Symbol: Serpine1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012620.1 | NP_036752.1 | 6 - 7 | 1113 | 71 | |
GenBank mRNA | M24067.1 | - | 6 - 7 | 1113 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
