Assay Details
Gene Symbol: Gnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031037.2 | NP_112299.1 | 7 - 8 | 922 | 67 | |
GenBank mRNA | AF277892.1 | - | 6 - 7 | 703 | 67 | |
AF397193.1 | - | 3 - 4 | 502 | 67 | ||
BC065579.1 | - | 7 - 8 | 922 | 67 | ||
DQ120485.1 | - | 6 - 7 | 687 | 67 | ||
DQ120486.1 | - | 6 - 7 | 687 | 67 | ||
FQ209560.1 | - | 3 - 4 | 310 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
