Assay Details
Gene Symbol: Dtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249158.1 | XP_006249220.1 | 1 - 2 | 166 | 83 | |
XM_006249159.1 | XP_006249221.1 | 1 - 2 | 166 | 83 | ||
GenBank mRNA | BC166738.1 | - | 1 - 2 | 107 | 83 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC166738.1, XM_006249158.1, XM_006249159.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
