Assay Details
Gene Symbol: Dtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107157.1 | NP_001100627.1 | 3 - 4 | 375 | 121 | |
XM_006249158.1 | XP_006249220.1 | 2 - 3 | 321 | 121 | ||
XM_006249159.1 | XP_006249221.1 | 2 - 3 | 321 | 121 | ||
XM_008769137.1 | XP_008767359.1 | 2 - 3 | 176 | 121 | ||
GenBank mRNA | AY724523.1 | - | 1 - 2 | 156 | 121 | |
BC166738.1 | - | 2 - 3 | 262 | 121 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC166738.1, NM_001107157.1, XM_006249158.1, XM_006249159.1, XM_008769137.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
