Assay Details
Gene Symbol: Tinf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006962.1 | NP_001006963.1 | 2 - 3 | 530 | 85 | |
XM_006251940.2 | XP_006252002.1 | 2 - 3 | 1518 | 85 | ||
XM_006251942.2 | XP_006252004.1 | 2 - 3 | 1518 | 85 | ||
XM_006251943.2 | XP_006252005.1 | 2 - 3 | 1519 | 85 | ||
XM_006251944.2 | XP_006252006.1 | 2 - 3 | 1518 | 85 | ||
GenBank mRNA | BC083766.1 | - | 2 - 3 | 530 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
