Assay Details
Gene Symbol: Gsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053906.2 | NP_446358.2 | 8 - 9 | 598 | 111 | |
GenBank mRNA | CK357934.1 | - | 1 - 2 | 61 | 111 | |
U73174.1 | - | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
