Assay Details
Gene Symbol: Grtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031813.2 | NP_001026983.1 | 1 - 2 | 119 | 79 | |
NM_001170477.1 | NP_001163948.1 | 1 - 2 | 122 | 79 | ||
GenBank mRNA | BC097985.1 | - | 1 - 2 | 73 | 79 | |
CB315310.1 | - | 1 - 2 | 122 | 79 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC097985.1, NM_001031813.2, NM_001170477.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
