Assay Details
Gene Symbol: Grtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031813.2 | NP_001026983.1 | 5 - 6 | 674 | 73 | |
NM_001170477.1 | NP_001163948.1 | 6 - 7 | 836 | 73 | ||
GenBank mRNA | BC097985.1 | - | 5 - 6 | 628 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
