Assay Details
Gene Symbol: Dlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127446.1 | NP_001120918.1 | 13 - 14 | 3447 | 137 | |
GenBank mRNA | D31962.1 | - | 13 - 14 | 3447 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
heart morphogenesis
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
cytoskeleton organization
signal transduction
activation of phospholipase C activity
Rho protein signal transduction
negative regulation of cell population proliferation
regulation of cell shape
hindbrain morphogenesis
actin cytoskeleton organization
negative regulation of cell migration
forebrain development
cellular response to insulin stimulus
regulation of actin cytoskeleton organization
negative regulation of Rho protein signal transduction
positive regulation of protein dephosphorylation
focal adhesion assembly
negative regulation of stress fiber assembly
negative regulation of focal adhesion assembly
positive regulation of execution phase of apoptosis
GTPase activator activity
lipid binding
vinculin binding
SH2 domain binding
phospholipase binding
neural tube closure
heart morphogenesis
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
cytoskeleton organization
signal transduction
activation of phospholipase C activity
Rho protein signal transduction
negative regulation of cell population proliferation
regulation of cell shape
hindbrain morphogenesis
actin cytoskeleton organization
negative regulation of cell migration
forebrain development
cellular response to insulin stimulus
regulation of actin cytoskeleton organization
negative regulation of Rho protein signal transduction
positive regulation of protein dephosphorylation
focal adhesion assembly
negative regulation of stress fiber assembly
negative regulation of focal adhesion assembly
positive regulation of execution phase of apoptosis
GTPase activator activity
lipid binding
vinculin binding
SH2 domain binding
phospholipase binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P145087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19886 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P145087 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P145087 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P145087 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P309034 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P821587 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10788472 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788464 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788470 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788467 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788474 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788468 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788469 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788471 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788466 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788476 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788473 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788475 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788465 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788463 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10788462 | - | - |
Rat Genome 230 2.0 Array | 1389894_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373291_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389894_at | - | - |
Rat Genome 230 2.0 Array | 1382715_at | - | - |
Rat Genome 230 2.0 Array | 1373291_at | - | - |
Rat Genome 230 2.0 Array | 1392520_PM_at | - | - |
Rat Genome 230 2.0 Array | 1382715_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392520_at | - | - |
Affymetrix HT Rat Focus Array | 1392520_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389894_at | - | - |
Affymetrix HT Rat Focus Array | 1389894_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392520_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1600015989.rn.1 | TC1600002121.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1600015986.rn.1 | TC1600002121.rn.1 | RTA TC |
Rat Expression Set 230 | 1389894_at | - | - |
Rat Expression Set 230 | 1373291_PM_at | - | - |
Rat Expression Set 230 | 1389894_PM_at | - | - |
Rat Expression Set 230 | 1373291_at | - | - |
Rat Expression Set 230 | 1382715_at | - | - |
Rat Expression Set 230 | 1392520_at | - | - |
Rat Expression Set 230 | 1382715_PM_at | - | - |
Rat Expression Set 230 | 1392520_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788462 | - | - |
Rat Genome U34 Set | D31962cds_at | - | - |
Rat Genome U34 Set | D31962UTR#1_at | - | - |
Rat Genome U34 Set | rc_AA850419_at | - | - |
Rat Genome U34 Set | rc_AI011286_at | - | - |
Rat Genome U34 Set | rc_AA818382_at | - | - |
Rat Genome U34 Set | rc_AI176713_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360349 | - | - |
