Assay Details
Gene Symbol: Dlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127446.1 | NP_001120918.1 | 1 - 2 | 322 | 78 | |
GenBank mRNA | D31962.1 | - | 1 - 2 | 322 | 78 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the D31962.1, NM_001127446.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
