Assay Details
Gene Symbol: Dlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127446.1 | NP_001120918.1 | 7 - 8 | 2307 | 63 | |
GenBank mRNA | D31962.1 | - | 7 - 8 | 2307 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
