Assay Details
Gene Symbol: Mak16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014002.1 | NP_001014024.1 | 3 - 4 | 219 | 77 | |
GenBank mRNA | BC090035.1 | - | 3 - 4 | 219 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
