Assay Details
Gene Symbol: Dctn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106085.1 | NP_001099555.1 | 6 - 7 | 557 | 118 | |
GenBank mRNA | FQ212035.1 | - | 6 - 7 | 543 | 118 | |
FQ226689.1 | - | 6 - 7 | 580 | 118 | ||
FQ233791.1 | - | 6 - 7 | 574 | 118 | ||
FQ234632.1 | - | 6 - 7 | 580 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
