Assay Details
Gene Symbol: Dctn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106085.1 | NP_001099555.1 | 1 - 2 | 106 | 81 | |
GenBank mRNA | FQ212035.1 | - | 1 - 2 | 92 | 81 | |
FQ226689.1 | - | 1 - 2 | 129 | 81 | ||
FQ233791.1 | - | 1 - 2 | 123 | 81 | ||
FQ234632.1 | - | 1 - 2 | 129 | 81 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001106085.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
