Assay Details
Gene Symbol: Agpat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134744.1 | NP_001128216.1 | 3 - 4 | 398 | 125 | |
XM_006253381.2 | XP_006253443.1 | 3 - 4 | 670 | 125 | ||
GenBank mRNA | BC168222.1 | - | 3 - 4 | 398 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
