Assay Details
Gene Symbol: Ptpn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145683.1 | NP_663716.1 | 2 - 3 | 232 | 71 | |
XM_006249820.2 | XP_006249882.1 | 2 - 3 | 707 | 71 | ||
XM_006249821.2 | XP_006249883.1 | 2 - 3 | 362 | 71 | ||
GenBank mRNA | U28356.1 | - | 2 - 3 | 232 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
