Assay Details
Gene Symbol: Uchl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012149.1 | NP_001012149.1 | 1 - 2 | 158 | 112 | |
XM_006249960.2 | XP_006250022.1 | 1 - 2 | 193 | 112 | ||
XM_006249961.2 | XP_006250023.1 | 1 - 2 | 193 | 112 | ||
XM_006249962.2 | XP_006250024.1 | 1 - 2 | 194 | 112 | ||
XM_006249963.2 | XP_006250025.1 | 1 - 2 | 195 | 112 | ||
GenBank mRNA | BC088841.1 | - | 1 - 2 | 158 | 112 | |
FQ220301.1 | - | 1 - 2 | 164 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
