Assay Details
Gene Symbol: Uchl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012149.1 | NP_001012149.1 | 8 - 9 | 815 | 103 | |
XM_006249960.2 | XP_006250022.1 | 8 - 9 | 850 | 103 | ||
XM_006249962.2 | XP_006250024.1 | 8 - 9 | 851 | 103 | ||
GenBank mRNA | BC088841.1 | - | 8 - 9 | 815 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
ubiquitin-dependent protein catabolic process
biological_process
negative regulation of endopeptidase activity
protein deubiquitination
lateral ventricle development
midbrain development
forebrain morphogenesis
regulation of proteasomal protein catabolic process
molecular_function
RNA binding
thiol-dependent deubiquitinase
endopeptidase inhibitor activity
omega peptidase activity
proteasome binding
ubiquitin-dependent protein catabolic process
biological_process
negative regulation of endopeptidase activity
protein deubiquitination
lateral ventricle development
midbrain development
forebrain morphogenesis
regulation of proteasomal protein catabolic process
molecular_function
RNA binding
thiol-dependent deubiquitinase
endopeptidase inhibitor activity
omega peptidase activity
proteasome binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1060312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8075 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1060312 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1060312 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1060312 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17377 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1060312 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 61325 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 81930 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3671 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96659 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 67288 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7258 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6832 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37326 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 5770 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77772 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44428 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36033 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89093 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65803 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63453 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42183 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94470 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69853 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15229 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10764512 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764516 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764521 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764510 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764517 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764519 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764513 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764515 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764520 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764509 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764511 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764518 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764514 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10764508 | - | - |
Rat Genome 230 2.0 Array | 1383477_at | - | - |
Rat Genome 230 2.0 Array | 1383477_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383477_at | - | - |
Affymetrix HT Rat Focus Array | 1383477_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300002951.rn.1 | TC1300000421.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300002955.rn.1 | TC1300000421.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300002947.rn.1 | TC1300000421.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300002948.rn.1 | TC1300000421.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300002953.rn.1 | TC1300000421.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300002956.rn.1 | TC1300000421.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300002950.rn.1 | TC1300000421.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300002961.rn.1 | TC1300000421.rn.1 | RTA TC |
Rat Expression Set 230 | 1383477_PM_at | - | - |
Rat Expression Set 230 | 1383477_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764508 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21826476 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375610 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376954 | - | - |
