Assay Details
Gene Symbol: Ccnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107171.1 | NP_001100641.1 | 1 - 2 | 195 | 113 | |
XM_006249686.2 | XP_006249748.1 | 1 - 2 | 203 | 113 | ||
XM_008769457.1 | XP_008767679.1 | 1 - 2 | 203 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
