Assay Details
Gene Symbol: Ccnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107171.1 | NP_001100641.1 | 8 - 9 | 805 | 112 | |
XM_006249686.2 | XP_006249748.1 | 8 - 9 | 813 | 112 | ||
XM_006249687.2 | XP_006249749.1 | 4 - 5 | 3514 | 112 | ||
XM_008769457.1 | XP_008767679.1 | 7 - 8 | 684 | 112 | ||
XM_008769458.1 | XP_008767680.1 | 5 - 6 | 2590 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P079275 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P233147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74598 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P233147 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P079275 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P079275 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P233147 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P079275 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P233147 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P233147 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P323558 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 13628 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83032 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9413 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24470 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100458 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10767286 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767288 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767281 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767289 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767285 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767283 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767287 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767284 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767282 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10767280 | - | - |
Rat Genome 230 2.0 Array | 1385697_at | - | - |
Rat Genome 230 2.0 Array | 1385697_PM_at | - | - |
Rat Genome 230 2.0 Array | 1399100_at | - | - |
Rat Genome 230 2.0 Array | 1399100_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390470_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390470_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300001004.rn.1 | TC1300000195.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300001005.rn.1 | TC1300000195.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300001003.rn.1 | TC1300000195.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300001008.rn.1 | TC1300000195.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300001006.rn.1 | TC1300000195.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300001007.rn.1 | TC1300000195.rn.1 | RTA TC |
Rat Expression Set 230 | 1399100_PM_at | - | - |
Rat Expression Set 230 | 1385697_PM_at | - | - |
Rat Expression Set 230 | 1385697_at | - | - |
Rat Expression Set 230 | 1390470_PM_at | - | - |
Rat Expression Set 230 | 1399100_at | - | - |
Rat Expression Set 230 | 1390470_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767280 | - | - |
Rat Genome U34 Set | rc_AA996445_at | - | - |
Rat Genome U34 Set | rc_AA851251_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21291776 | - | - |
ABI Rat Genome Survey Microarray | 22151127 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359160 | - | - |
