Assay Details
Gene Symbol: Cd247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205304.1 | NP_001192233.1 | 5 - 6 | 432 | 77 | |
NM_170789.2 | NP_740770.2 | 6 - 7 | 513 | 77 | ||
XM_006250147.2 | XP_006250209.1 | 6 - 7 | 553 | 77 | ||
GenBank mRNA | BC097933.1 | - | 6 - 7 | 456 | 77 | |
CO562119.1 | - | 5 - 6 | 366 | 77 | ||
D13555.1 | - | 6 - 7 | 513 | 77 | ||
L08447.1 | - | 6 - 7 | 440 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
