Assay Details
Gene Symbol: Cd247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205304.1 | NP_001192233.1 | 6 - 7 | 474 | 106 | |
NM_170789.2 | NP_740770.2 | 7 - 8 | 555 | 106 | ||
XM_006250147.2 | XP_006250209.1 | 7 - 8 | 595 | 106 | ||
GenBank mRNA | BC097933.1 | - | 7 - 8 | 498 | 106 | |
CO562119.1 | - | 6 - 7 | 408 | 106 | ||
D13555.1 | - | 7 - 8 | 555 | 106 | ||
L08447.1 | - | 7 - 8 | 482 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P116899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108970 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P116899 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P116899 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P116899 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P463805 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 21448 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1991 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91360 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24040 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49138 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34208 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10765340 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765343 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765346 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765348 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765347 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765342 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765341 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765344 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765345 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10765339 | - | - |
Rat Genome 230 2.0 Array | 1388026_at | - | - |
Rat Genome 230 2.0 Array | 1370621_at | - | - |
Rat Genome 230 2.0 Array | 1371273_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371273_at | - | - |
Rat Genome 230 2.0 Array | 1370621_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388026_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388026_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388026_at | - | - |
Affymetrix HT Rat Focus Array | 1370621_at | - | - |
Affymetrix HT Rat Focus Array | 1370621_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300004770.rn.1 | TC1300000677.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300004771.rn.1 | TC1300000677.rn.1 | RTA TC |
Rat Expression Set 230 | 1388026_PM_at | - | - |
Rat Expression Set 230 | 1370621_at | - | - |
Rat Expression Set 230 | 1371273_PM_at | - | - |
Rat Expression Set 230 | 1371273_at | - | - |
Rat Expression Set 230 | 1388026_at | - | - |
Rat Expression Set 230 | 1370621_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765339 | - | - |
Rat Genome U34 Set | D13555_at | - | - |
Rat Genome U34 Set | D13556exon_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21586806 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370659 | - | - |
RatRef-12 Expression BeadChip | ILMN_49243 | - | - |
