Assay Details
Gene Symbol: Rassf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019365.3 | NP_062238.1 | 3 - 4 | 802 | 95 | |
XM_006249796.2 | XP_006249858.1 | 3 - 4 | 1663 | 95 | ||
XM_008769494.1 | XP_008767716.1 | 2 - 3 | 481 | 95 | ||
GenBank mRNA | AF002251.1 | - | 3 - 4 | 802 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
