Assay Details
Gene Symbol: Rassf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019365.3 | NP_062238.1 | 4 - 5 | 1100 | 132 | |
XM_006249796.2 | XP_006249858.1 | 4 - 5 | 1961 | 132 | ||
XM_008769494.1 | XP_008767716.1 | 3 - 4 | 779 | 132 | ||
GenBank mRNA | AF002251.1 | - | 4 - 5 | 1100 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
