Assay Details
Gene Symbol: Rxrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031765.1 | NP_113953.1 | 8 - 9 | 1359 | 126 | |
XM_006250212.2 | XP_006250274.1 | 7 - 8 | 1503 | 126 | ||
GenBank mRNA | AF016387.1 | - | 1 - 2 | 63 | 126 | |
AJ223083.1 | - | 7 - 8 | 894 | 126 | ||
BC091363.1 | - | 8 - 9 | 1359 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
transcription by RNA polymerase II
peripheral nervous system development
heart development
skeletal muscle tissue development
response to organic cyclic compound
neuron differentiation
regulation of myelination
response to retinoic acid
cellular response to hormone stimulus
steroid hormone mediated signaling pathway
positive regulation of transcription by RNA polymerase II
retinoic acid receptor signaling pathway
protein homotetramerization
positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
RNA polymerase II transcription regulatory region sequence-specific DNA binding
steroid hormone receptor activity
zinc ion binding
sequence-specific DNA binding
transcription, DNA-templated
transcription by RNA polymerase II
peripheral nervous system development
heart development
skeletal muscle tissue development
response to organic cyclic compound
neuron differentiation
regulation of myelination
response to retinoic acid
cellular response to hormone stimulus
steroid hormone mediated signaling pathway
positive regulation of transcription by RNA polymerase II
retinoic acid receptor signaling pathway
protein homotetramerization
positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
RNA polymerase II transcription regulatory region sequence-specific DNA binding
steroid hormone receptor activity
zinc ion binding
sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P027625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46097 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P027625 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P027625 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P027625 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P328016 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P269316 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700934179 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45127 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24708 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9622 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82350 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700934179 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10765420 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765418 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765416 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765419 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765414 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765415 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765423 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765421 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765417 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765422 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10765413 | - | - |
Rat Genome 230 2.0 Array | 1392271_PM_at | - | - |
Rat Genome 230 2.0 Array | 1370981_at | - | - |
Rat Genome 230 2.0 Array | 1370981_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392271_at | - | - |
Affymetrix HT Rat Focus Array | 1370981_at | - | - |
Affymetrix HT Rat Focus Array | 1370981_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392271_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392271_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300004983.rn.1 | TC1300000711.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300004985.rn.1 | TC1300000711.rn.1 | RTA TC |
Rat Expression Set 230 | 1370981_at | - | - |
Rat Expression Set 230 | 1370981_PM_at | - | - |
Rat Expression Set 230 | 1392271_at | - | - |
Rat Expression Set 230 | 1392271_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10765413 | - | - |
Rat Genome U34 Set | AF016387_at | - | - |
Rat Genome U34 Set | AF016387_g_at | - | - |
Rat Genome U34 Set | AJ223083_at | - | - |
Rat Genome U34 Set | rc_AA955071_i_at | - | - |
Rat Genome U34 Set | AJ223083_at | - | - |
Rat Genome U34 Set | AJ223083_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21337681 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371942 | - | - |
