Assay Details
Gene Symbol: Rnasel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182673.2 | NP_872614.1 | 3 - 4 | 1806 | 61 | |
XM_006249998.2 | XP_006250060.1 | 5 - 6 | 2193 | 61 | ||
XM_006249999.2 | XP_006250061.1 | 5 - 6 | 2397 | 61 | ||
XM_006250000.2 | XP_006250062.1 | 5 - 6 | 2483 | 61 | ||
XM_006250001.2 | XP_006250063.1 | 5 - 6 | 2172 | 61 | ||
XM_006250002.2 | XP_006250064.1 | 4 - 5 | 2248 | 61 | ||
XM_008769653.1 | XP_008767875.1 | 4 - 5 | 2760 | 61 | ||
GenBank mRNA | AY262823.1 | - | 3 - 4 | 1808 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
