Assay Details
Gene Symbol: Rnasel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182673.2 | NP_872614.1 | 4 - 5 | 1948 | 118 | |
XM_006249998.2 | XP_006250060.1 | 6 - 7 | 2335 | 118 | ||
XM_006249999.2 | XP_006250061.1 | 6 - 7 | 2539 | 118 | ||
XM_006250000.2 | XP_006250062.1 | 6 - 7 | 2625 | 118 | ||
XM_006250001.2 | XP_006250063.1 | 6 - 7 | 2314 | 118 | ||
XM_006250002.2 | XP_006250064.1 | 5 - 6 | 2390 | 118 | ||
XM_006250004.2 | XP_006250066.1 | 5 - 6 | 2931 | 118 | ||
XM_008769653.1 | XP_008767875.1 | 5 - 6 | 2902 | 118 | ||
GenBank mRNA | AY262823.1 | - | 4 - 5 | 1950 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
