Assay Details
Gene Symbol: Hsd17b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017235.3 | NP_058931.1 | 5 - 6 | 714 | 74 | |
XM_006250215.2 | XP_006250277.1 | 5 - 6 | 737 | 74 | ||
XM_006250216.2 | XP_006250278.1 | 5 - 6 | 737 | 74 | ||
XM_006250217.2 | XP_006250279.1 | 5 - 6 | 737 | 74 | ||
GenBank mRNA | FM073997.1 | - | 5 - 6 | 73 | ||
FN803430.1 | - | 3 - 4 | 327 | 74 | ||
FQ113345.1 | - | 5 - 6 | 714 | 74 | ||
U44803.1 | - | 5 - 6 | 659 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
