Assay Details
Gene Symbol: Tor1aip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145092.2 | NP_659560.2 | 3 - 4 | 805 | 132 | |
XM_006250027.2 | XP_006250089.1 | 3 - 4 | 350 | 132 | ||
XM_006250028.2 | XP_006250090.1 | 3 - 4 | 936 | 132 | ||
GenBank mRNA | BC086987.1 | - | 3 - 4 | 805 | 132 | |
U20286.1 | - | 3 - 4 | 262 | 132 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_006250027.2, XM_006250028.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
