Assay Details
Gene Symbol: Il10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012854.2 | NP_036986.2 | 1 - 2 | 165 | 74 | |
XM_006249712.2 | XP_006249774.1 | 1 - 2 | 504 | 74 | ||
XM_008769426.1 | XP_008767648.1 | 1 - 2 | 504 | 74 | ||
GenBank mRNA | L02926.1 | - | 1 - 2 | 165 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
