Assay Details
Gene Symbol: Pak6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106498.1 | NP_001099968.1 | 9 - 10 | 2380 | 85 | |
XM_008762091.1 | XP_008760313.1 | 8 - 9 | 3680 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
