Assay Details
Gene Symbol: Mid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191889.1 | NP_001178818.1 | 1 - 2 | 166 | 116 | |
XM_006234247.2 | XP_006234309.1 | 1 - 2 | 551 | 116 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001191889.1, XM_006234247.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
