Assay Details
Gene Symbol: Mid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191889.1 | NP_001178818.1 | 6 - 7 | 1368 | 152 | |
XM_006234247.2 | XP_006234309.1 | 6 - 7 | 1753 | 152 | ||
XM_006234248.2 | XP_006234310.1 | 6 - 7 | 1309 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
