Assay Details
Gene Symbol: Ndufs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106489.1 | NP_001099959.1 | 2 - 3 | 316 | 82 | |
GenBank mRNA | FQ212516.1 | - | 2 - 3 | 140 | 82 | |
FQ219879.1 | - | 2 - 3 | 143 | 82 | ||
FQ229748.1 | - | 2 - 3 | 163 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
