Assay Details
Gene Symbol: Nfe2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234396.2 | XP_006234458.1 | 3 - 4 | 618 | 65 | |
XM_006234397.2 | XP_006234459.1 | 3 - 4 | 563 | 65 | ||
XM_006234398.2 | XP_006234460.1 | 3 - 4 | 1170 | 65 | ||
GenBank mRNA | BC061724.1 | - | 3 - 4 | 485 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
