Assay Details
Gene Symbol: Cd82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031797.2 | NP_113985.2 | 4 - 5 | 388 | 56 | |
XM_006234597.2 | XP_006234659.1 | 4 - 5 | 462 | 56 | ||
XM_006234598.1 | XP_006234660.1 | 4 - 5 | 276 | 56 | ||
XM_008762039.1 | XP_008760261.1 | 3 - 4 | 181 | 56 | ||
GenBank mRNA | BC072498.1 | - | 4 - 5 | 388 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
