Assay Details
Gene Symbol: Cd82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031797.2 | NP_113985.2 | 6 - 7 | 591 | 89 | |
XM_006234597.2 | XP_006234659.1 | 6 - 7 | 665 | 89 | ||
XM_006234598.1 | XP_006234660.1 | 6 - 7 | 479 | 89 | ||
XM_008762039.1 | XP_008760261.1 | 5 - 6 | 384 | 89 | ||
GenBank mRNA | BC072498.1 | - | 6 - 7 | 591 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
