Assay Details
Gene Symbol: Arhgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107747.2 | NP_001101217.2 | 3 - 4 | 364 | 80 | |
XM_006234551.1 | XP_006234613.1 | 4 - 5 | 401 | 80 | ||
XM_006234552.2 | XP_006234614.1 | 4 - 5 | 431 | 80 | ||
XM_006234553.2 | XP_006234615.1 | 4 - 5 | 401 | 80 | ||
XM_006234554.1 | XP_006234616.1 | 5 - 6 | 443 | 80 | ||
XM_006234556.1 | XP_006234618.1 | 4 - 5 | 378 | 80 | ||
XM_006234557.2 | XP_006234619.1 | 4 - 5 | 408 | 80 | ||
XM_006234558.2 | XP_006234620.1 | 3 - 4 | 349 | 80 | ||
XM_006234560.2 | XP_006234622.1 | 3 - 4 | 390 | 80 | ||
GenBank mRNA | CK480577.1 | - | 3 - 4 | 364 | 80 | |
FQ224002.1 | - | 3 - 4 | 324 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
