Assay Details
Gene Symbol: Idh3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053581.1 | NP_446033.1 | 8 - 9 | 782 | 109 | |
XM_006235048.2 | XP_006235110.1 | 8 - 9 | 819 | 109 | ||
GenBank mRNA | BC079113.1 | - | 8 - 9 | 782 | 109 | |
FQ214820.1 | - | 8 - 9 | 780 | 109 | ||
FQ215180.1 | - | 8 - 9 | 786 | 109 | ||
FQ215350.1 | - | 8 - 9 | 782 | 109 | ||
FQ217667.1 | - | 2 - 3 | 140 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
