Assay Details
Gene Symbol: Cry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133405.2 | NP_596896.2 | 9 - 10 | 1551 | 73 | |
GenBank mRNA | BC166416.1 | - | 9 - 10 | 1551 | 73 | |
FQ211347.1 | - | 9 - 10 | 1561 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
