Assay Details
Gene Symbol: Polr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031773.1 | NP_113961.1 | 11 - 12 | 1974 | 62 | |
GenBank mRNA | AF025424.1 | - | 11 - 12 | 1974 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
