Assay Details
Gene Symbol: Slc39a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039196.1 | NP_001034285.1 | 2 - 3 | 475 | 93 | |
XM_006234501.2 | XP_006234563.1 | 2 - 3 | 681 | 93 | ||
XM_006234502.2 | XP_006234564.1 | 2 - 3 | 685 | 93 | ||
GenBank mRNA | BC112321.1 | - | 2 - 3 | 475 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
