Assay Details
Gene Symbol: Srp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106497.1 | NP_001099967.1 | 2 - 3 | 181 | 66 | |
GenBank mRNA | BC166931.1 | - | 2 - 3 | 137 | 66 | |
FQ210704.1 | - | 1 - 2 | 81 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
