Assay Details
Gene Symbol: Rmdn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014046.2 | NP_001014068.1 | 3 - 4 | 470 | 92 | |
GenBank mRNA | BC082081.1 | - | 3 - 4 | 432 | 92 | |
DN931790.1 | - | 3 - 4 | 502 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
