Assay Details
Gene Symbol: Rmdn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014046.2 | NP_001014068.1 | 4 - 5 | 617 | 107 | |
GenBank mRNA | BC082081.1 | - | 4 - 5 | 579 | 107 | |
DN931790.1 | - | 4 - 5 | 649 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
