Assay Details
Gene Symbol: Mtmr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108123.1 | NP_001101593.1 | 7 - 8 | 1648 | 109 | |
XM_006242521.2 | XP_006242583.1 | 8 - 9 | 1958 | 109 | ||
XM_006242522.1 | XP_006242584.1 | 7 - 8 | 775 | 109 | ||
XM_008765932.1 | XP_008764154.1 | 8 - 9 | 665 | 109 | ||
GenBank mRNA | FQ217011.1 | - | 8 - 9 | 842 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
